Anti-IDH1 Mouse Monoclonal Antibody (CF594) [clone: IDH1/1152]

Proveedor: Biotium
BNC941152-500 BNC941152-100
BNC941152-500EA 560 EUR
BNC941152-500 BNC941152-100
Anti-IDH1 Mouse Monoclonal Antibody (CF594) [clone: IDH1/1152]
Anticuerpos
This antibody recognizes a 45 kDa protein, which is identified as isocitrate dehydrogenase (IDH1). It belongs to the isocitrate and isopropylmalate dehydrogenases family. IDH1 catalyzes the third step of the citric acid cycle, which involves the oxidative decarboxylation of isocitrate, formingalpha-ketoglutarate and CO2 in a two-step reaction. The first step involves the oxidation of isocitrate to the intermediate oxalosuccinate, while the second step involves the production ofalpha-ketoglutarate. During this process, either NADH or NADPH is produced along with CO2. Recently, an inactivating mutation of IDH1 has been implicated in glioblastoma. IDH1 appears to function as a tumor suppressor that, when mutationally inactivated, contributes to tumorigenesis in part through induction of the HIF-1 pathway.

Type: Primary
Antigen: IDH1
Clonality: Monoclonal
Clone: IDH1/1152
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR