Anti-PTH Mouse Monoclonal Antibody (Purified) [clone: PTH/1174]
Proveedor: Biotium
BNUB1174-500 BNUB1174-100
BTIUBNUB1174-500EA 511 EUR
BTIUBNUB1174-500 BTIUBNUB1174-100
Anti-PTH Mouse Monoclonal Antibody (Purified) [clone: PTH/1174]
Anticuerpos
The epitope of this MAb maps in the C-terminus of PTH, a hormone produced by the parathyroid gland that regulates the concentration of calcium and phosphorus in extracellular fluid. This hormone elevates blood Ca2 levels by dissolving the salts in bone and preventing their renal excretion.It is produced in the parathyroid gland as an 84 amino acid single chain polypeptide. It can also be secreted as N-terminal truncated fragments or C-terminal fragments after intracellular degradation, as in case of hypercalcemia. Defects in this gene are a cause of familial isolated hypoparathyroidism (FIH);More Product Information