Anti-BrdU Mouse Monoclonal Antibody (Purified) [clone: 85-2C8]
Proveedor: Biotium
BNUB0886-500 BNUB0886-100
BNUB0886-500EA 511 EUR
BNUB0886-500 BNUB0886-100
Anti-BrdU Mouse Monoclonal Antibody (Purified) [clone: 85-2C8]
Anticuerpos
This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.
Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: 85-2C8
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species