Analytical HPLC columns, ultra C4 (USP L26)
Proveedor: Restek
REST9102552-700EA 583 EUR
REST9102552-700 REST9102535-700 REST910253E REST910257E REST9102315-700 REST910233E REST9102365-700 REST9102553-700 REST9102535 REST9102332-700 REST9102332 REST9102333 REST9102575 REST9102531 REST9102532 REST9102533 REST9102335 REST9102571 REST9102572 REST9102331 REST9102573 REST910252E REST9102333-700 REST910236E REST9102363-700 REST910256E REST9102555-700 REST9102512-700 REST9102525-700 REST9102523 REST9102572-700 REST9102525 REST9102365 REST9102563 REST9102565 REST9102521 REST9102522 REST9102361 REST9102362 REST9102561 REST9102363 REST9102562 REST9102522-700 REST910231E REST910250210 REST910250212 REST910251E REST910235E REST9102573-700 REST910255E REST9102523-700 REST9102352-700 REST9102335-700 REST9102362-700 REST9102512 REST9102513 REST9102315 REST9102515 REST9102552 REST9102355 REST9102553 REST9102311 REST9102513-700 REST9102312 REST9102511 REST9102555 REST9102313 REST9102353-700 REST9102351 REST9102352 REST9102565-700 REST9102551 REST9102353 REST9102355-700 REST9102533-700 REST9102562-700 REST9102575-700 REST9102313-700 REST9102515-700 REST9102532-700 REST9102563-700 REST9102312-700
Analytical HPLC columns, ultra C4 (USP L26)
Columnas de cromatografía
Ultra C4 columns made of high-purity, type-B silica are designed for selective and reliable HPLC applications.
  • Exceptionally stable C4 packaging
  • High bonding coverage and base deactivation
  • Less retention than C18 or C8 phases
  • Minimises activity and creates high-density bonding