Deactivated fused silica tubing (Nonpolar)
Proveedor: Restek
10009 10010 10011 10020 10021 10030 10031
REST10009EA 237 EUR
REST10009 REST10010 REST10011 REST10020 REST10021 REST10030 REST10031
Deactivated fused silica tubing (Nonpolar)
Columnas de cromatografía
This fused silica tubing is commonly used as guard columns, transfer lines or for long retention gaps.
  • Inertness tested with the grob test mix
  • Maximum temperature: 325 °C
  • Lengths of tubings are nonpolar and provided in 5-meter banded sections