Indicating oxygen traps
Proveedor: Restek
22010 22011
REST22010EA 210 EUR
REST22010 REST22011
Indicating oxygen traps
Filtros Filtros de gas
These traps have a heavy-walled with glass body, protected by PC sleeve, which prevents oxygen and water infusion. Indicator changes from light green to grey as oxygen is trapped.
  • Prepurged for fast stabilisation
  • Maximum operating pressure 100 psi
  • Reduces oxygen to 0,1 ppm
  • 10 μm frits at inlet and outlet
  • Optimal flow rate: <150 ml/min