Capillary columns, Rtx®-Dioxin2 (fused silica)
Proveedor: Restek
10759 10758
REST10759EA 1148 EUR
REST10759 REST10758
Capillary columns, Rtx®-Dioxin2 (fused silica)
Columnas de cromatografía
These capillary columns are isomer specificity for 2,3,7,8-TCDD and 2,3,7,8-TCDF achieved with one GC column. Unique selectivity for toxic dioxin and furan congeners which allows to use as a confirmation GC column.
  • Thermally stable to 340 °C for longer lifetime