Septum packed purge port weldment, Siltek®
Proveedor: Restek
21685 21691
REST21685EA 24 EUR
REST21685 REST21691
Septum packed purge port weldment, Siltek®
Suministros de cromatografía Piezas de repuesto para cromatografía
This Siltek® treated septum packed purge port weldment for Agilent 5890 GC makes weldment inert and eliminates adsorption of sensitive compounds such as, DDT and Endrin.
Información para pedidos: Viton O-rings and appropriate septa have to be ordered separately.