GC ferrules, Graphite Vespel®
Proveedor: Restek
20420 20421 20422 20423
REST20420EA 83 EUR
REST20420 REST20421 REST20422 REST20423
GC ferrules, Graphite Vespel®
Racores y conectores
These ferrules can be used with Vacuum Vu-Union® connectors.
  • Made of 60% Vespel® and 40% graphite
  • Maximum operating temperature up to 400 °C