Preassembled vial kits
Proveedor: Restek
24630 24631 24632 24633 24634 24635 24636 24637
REST24630EA 70 EUR
REST24630 REST24631 REST24632 REST24633 REST24634 REST24635 REST24636 REST24637
Preassembled vial kits
Viales
Vial kits with 8 mm/425 screw-thread, unmarked vials, septa and caps (caps are attached to vials).