Caps for screw thread vials
Proveedor: Restek
21176 21177
REST21176EA 14 EUR
REST21176 REST21177
Caps for screw thread vials
Tapas Tapones para viales de cromatografía
Black PP, open hole caps fit all 2,0 ml, 12×32 mm, screw-thread 8 mm/425 vial-based autosamplers.