Rivaroxaban

Proveedor: Cayman Chemical

16043-10 16043-1 16043-50 16043-5
CAYM16043-10EA 174 EUR
CAYM16043-10 CAYM16043-1 CAYM16043-50 CAYM16043-5
Rivaroxaban
Rivaroxaban

Rivaroxaban is an orally active, direct inhibitor of Factor Xa (Ki = 0,4 nm), which is a crucial component of the intrinsic and extrinsic pathways of the blood coagulation cascade.


  • Purity: ≥98%


It demonstrates >10 000-fold greater selectivity for Factor Xa compared to other related serine proteases. In various animal arterial and venous thrombosis models, rivaroxaban is reported to inhibit thrombin formation without prolonging bleeding time and has been approved for clinical use as an anticoagulant in the prevention of stroke and the treatment of venous thromboembolisms.

Formula: C₁₉H₁₈ClN₃O₅S
Número MDL: MFCD11974010
Núm. CAS: 366789-02-8

Order Now

ESPECIFICACIONES

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR