O-rings, graphite
Proveedor: Restek
21475 21476
REST21475EA 71 EUR
REST21475 REST21476
O-rings, graphite
Juntas Anillos de Sellado
Graphite O-rings for PerkinElmer Auto SYS XL or Clarus GCs with PSS injector.
Max temp.: 450 °C