GC capillary column connectors, Swagelok®
Proveedor: Restek
22540 22541 22542 22543 22544 22545 22546 22547 22548 22549 22550 22551 22552 22553 22554 22555 22556 22557 22558 22559 22561 22562 22563 22564 22565 22566 22567 22568 22569 22570 22571 22573 22574 22575 22576 22577 22578 22579 22580 22581 22582 22583 22584 22585 22586 22587 22588 22589 22590 22591 22592 22593 22594 22595 22596 22597 22610 22611 22612 22613 22614 22615 22616 22617 22618 23156 23153 23110 23129 23111 23124 23122 23116 23150 23192 23170 23173 23160 23161 23159 23103 23106 23104 23107 23115 23121
REST22540EA 185 EUR
REST22540 REST22541 REST22542 REST22543 REST22544 REST22545 REST22546 REST22547 REST22548 REST22549 REST22550 REST22551 REST22552 REST22553 REST22554 REST22555 REST22556 REST22557 REST22558 REST22559 REST22561 REST22562 REST22563 REST22564 REST22565 REST22566 REST22567 REST22568 REST22569 REST22570 REST22571 REST22573 REST22574 REST22575 REST22576 REST22577 REST22578 REST22579 REST22580 REST22581 REST22582 REST22583 REST22584 REST22585 REST22586 REST22587 REST22588 REST22589 REST22590 REST22591 REST22592 REST22593 REST22594 REST22595 REST22596 REST22597 REST22610 REST22611 REST22612 REST22613 REST22614 REST22615 REST22616 REST22617 REST22618 REST23156 REST23153 REST23110 REST23129 REST23111 REST23124 REST23122 REST23116 REST23150 REST23192 REST23170 REST23173 REST23160 REST23161 REST23159 REST23103 REST23106 REST23104 REST23107 REST23115 REST23121
GC capillary column connectors, Swagelok®
Racores y conectores
Parker's fittings are ideal for installing new equipment, modifying existing instrumentation or replacing worn connections.
  • Fittings are available in brass or stainless steel
  • Treated with Siltek®/Sulfinert or Silcosteel®-CR