Fittings for HPLC and UHPLC, reusable, EXP®
Proveedor: Restek
25937 25938 25939 25926 25928 25930 25932 25933 25934 25935 25927 25929 25931 25936 25940
REST25937EA 79 EUR
REST25937 REST25938 REST25939 REST25926 REST25928 REST25930 REST25932 REST25933 REST25934 REST25935 REST25927 REST25929 REST25931 REST25936 REST25940
Fittings for HPLC and UHPLC, reusable, EXP®
Racores y conectores
EXP® reusable fittings for easy and reliable HPLC and UHPLC connections for 10 to 32 fittings and ¹/₁₆" tubing.
  • Hand-tight fitting style achieves effortless HPLC seals without any tools for a 600+ bar seal
  • Both hand-tight and hex-head styles wrench-tighten for reliable UHPLC use up to 1379+ bar
  • Ferrule can be installed repeatedly without compromising high-pressure seal
  • Hybrid design combines the durability of titanium with the sealing ability of PEEK
  • Cutting-edge system provides ZDV (zero dead volume) connection to any 10 to 32 female port
  • Compatible with ¹/₁₆" PEEK and stainless steel tubing