GraceResolv™ high-resolution flash cartridges
Proveedor: Alltech
5142034 8618522 8618944 8618943 8618942 8618941 5154065 8618520 8618509 8618502 5142051 5142050 5152371
557-0046EA 0 EUR
557-0046 557-0054 557-0061 557-0060 557-0059 557-0058 557-0053 557-0052 557-0051 557-0050 557-0049 557-0048 557-0047
GraceResolv™ high-resolution flash cartridges
Columnas de cromatografía
GraceResolv™ cartridges are packed with a grade of Davisil® silica designed to maximise performance of today's flash instruments.
  • High resolution
  • Increased loading capacity
  • Reproducible performance
  • Compatible with all flash instruments