Multi-channel pipettes, electronic, with bluetooth, E1-ClipTip™
Proveedor: Thermo Fisher Scientific
4671000BT 4671010BT 4671020BT 4671030BT 4671040BT 4671050BT 4671060BT 4671070BT 4671080BT 4671090BT 4671100BT
613-5752EA 1770 EUR
613-5752 613-5756 613-5760 613-5761 613-5753 613-5757 613-5762 613-5754 613-5758 613-5759 613-5755
Multi-channel pipettes, electronic, with bluetooth, E1-ClipTip™
Pipetas Micropipetas
With enhanced versatility and customisation, these pipettors automate daily 96- and 384-well microplate pipetting tasks. Electronic tip ejection and index finger pipetting action allow the thumb to relax during pipetting.
  • ClipTip Interlocking Technology: Tip is locked and sealed in place until ejected − no dropped tips that waste valuable samples
  • Two operating options for every application: Step-based Matrix or Pre-sets with settings for most common applications
  • Five user profiles, personalised user interface and password protection offer unified operation for multiple users and environments
  • Ability to store up to 30 common protocols with a specific name and calibration in the Programs function
  • Service and calibration trackers remind users when the pipette requires service or repair
  • Lithium-ion battery ensures long lasting performance; unit may be recharged by a charging stand or plug
  • Pipettes can be programmed via PC by connecting to the new My Pipette™ Creator app via Bluetooth* or USB connection
Información para pedidos: Supplied with universal power supply. 2 year extended warranty only available via web registration.
Precaución: Please note: ClipTip™ 384 and ClipTip™ 12,5 µl tips and compatible E1-ClipTip™ models do not have interlocking technology.
* Bluetooth models are not available in all countries.