Volumetric flasks, class A, PFA with screw cap, Corning®
Proveedor: Corning
5650P-250 5650P-100 5650P-10 5650P-25 5650P-500 5650P-50
CORN5650P-250EA 1645.32 EUR
CORN5650P-250 CORN5650P-100 CORN5650P-10 CORN5650P-25 CORN5650P-500 CORN5650P-50
Volumetric flasks, class A, PFA with screw cap, Corning®
Matraces Matraces aforados
Made of PFA, translucent, with PFA screw cap.
  • With an individually calibrated graduation line
  • Recommended routine cleaning up to 60 °C will preserve graduations
  • Exposure to temperatures above 60 °C may affect accuracy
These reusable flasks are manufactured to Class A tolerances as defined by ISO 1042.
The accuracy of volumetric ware depends on the care used in calibrating it, using the correct type of ware for the application, handling the ware properly and ensuring the ware is clean. Calibration of volumetric ware is usually done at 20 °C and theMore Product Information