HPLC columns, Poroshell 120 SB-C18
Proveedor: Agilent
683975-902 685775-902 681975-902 681975-302 683775-902 683975-302 685975-302 685975-902 687775-902 687975-302 687975-902 689775-902 689975-302 689975-902 681775-902
554-1733EA 910 EUR
554-1733 554-4337 HEWL681975-902 HEWL681975-302 554-4336 554-1734 554-1732 554-1731 HEWL687775-902 554-1730 554-1729 HEWL689775-902 HEWL689975-302 HEWL689975-902 HEWL681775-902
HPLC columns, Poroshell 120 SB-C18
Columnas de cromatografía
Poroshell 120 is made with a superficially porous particle that has a solid silica core and a porous outer layer. This particle provides high efficiency at lower pressures and is ideal for fast or high-resolution separations of many types of analytes.
  • Provides greater alternative selectivity
  • Steric protection of the bonded phase resists degradation
  • Ideal for basic, neutral, acidic samples or aggressive low pH mobile phases
The StableBond SB-C18 bonded phase is made by chemically bonding a sterically-protected C18 stationary phase to the porous shellMore Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Carga de carbono 8% 8% 8%
Extremo tapado No No No
Presión máx. 600 bar 600 bar 600 bar
Rango de pH 1,0 - 8,0 1,0 - 8,0 1,0 - 8,0
Surface area 130 m²/g 130 m²/g 130 m²/g
Temperature limit 90 °C 90 °C 90 °C
Código USP L1 L1 L1