Eyewash bottles, Duo
Proveedor: Plum Safety
4801 4800 4861
118-0094EA 172 EUR
118-0094 118-0093 118-0480
Eyewash bottles, Duo
Suministros de primeros auxilios Estaciones de lavado de ojos
Sterile 0,9 % Sodium chloride solution is ideal for injuries with foreign bodies like dust, dirt ect. Sterile 4,9 % Phosphate buffer solution is perfect for neutralising acids and alkalis.
  • Complete with an eye device for rinsing both eyes at the same time
  • Also for refilling of the Duo eye wash station
Rinsing time: ~2 min. / 5 min.
Shelf life: 3 years