Gel filtration chromatography columns, ZORBAX
Proveedor: Agilent
820675-111 820950-908 820950-911 884973-701 884973-901 884973-902 820212-911 877974-901 877974-910
HEWL820675-111EA 433 EUR
HEWL820675-111 HEWL820950-908 HEWL820950-911 HEWL884973-701 HEWL884973-901 HEWL884973-902 HEWL820212-911 HEWL877974-901 HEWL877974-910
Gel filtration chromatography columns, ZORBAX
Columnas de cromatografía
These ZORBAX GF-250 and GF-450 size exclusion (gel filtration) columns are ideal for the size separations of proteins and other biomolecules.
  • High efficiency and reproducibility with short analysis time
  • Hydrophilic diol bonded phase for good protein recovery
  • Compatible with organic modifiers and denaturants
  • Wide usable pH range (pH 3 to 8)
Some common applications includes separations of protein monomers, dimers and aggregates, desalting, protein molecular weight estimation and separations of modified proteins.