Filter vials, SINGLE StEP™ eXtreme
Proveedor: Restek
25874 25875 25876 25877 25878 25879 25880
REST25874EA 361 EUR
REST25874 REST25875 REST25876 REST25877 REST25878 REST25879 REST25880
Filter vials, SINGLE StEP™ eXtreme
Filtros Filtros sin jeringas
SINGLE StEP® eXtreme filter vials are recommended for samples containing less than 30% solid particulates and also offers fast sample filtration with a squeeze of the fingers. Fit most standard 12×32 mm autosamplers, including UHPLC instruments. It allow compounds to be separated from matrix, resulting in both higher signal-to-noise ratios and more differentiated peaks.
  • Minimise sample loss by eliminating multiple transfers
  • Colour coded caps allow easy identification of 0,2 μm or 0,45 μm membranes in PVDF, PTFE, PES or nylon
  • Pre-slit PTFE/silicone caps help eliminate broken autosampler needles and cored septa
  • Rugged PP vial houses insert with 450 μl loading capacity and low dead volume 120 μl