Organic reference standards, desogestrel standards, BP

Proveedor: LGC Standards PROMOCHEM

BP816 BP817
PROOBP816EA 256 EUR
PROOBP816 PROOBP817
Organic reference standards, desogestrel standards, BP
Normativas Estándares orgánicos

Order Now

ESPECIFICACIONES

Descripción Desogestrel impurity D Desogestrel impurity E
Environmentally Preferable
CAS No. 54048-10-1 70805-85-5
Formula C₂₂H₂₈O₂ C₂₂H₃₀O₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR