Risedronate Related Compounds, Reference Standard

Proveedor: USP

1604621 1604610 1604632 1604643
1604621.EA 1140 EUR
1604621. 1604610. 1604632. 1604643.
Risedronate Related Compounds, Reference Standard
Normativas Estándares orgánicos

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Certificaciones: US Pharmacopea Reference Standard

Order Now

ESPECIFICACIONES

Descripción Risedronate Related Compound A Risedronate Sodium Risedronate Related Compound B Risedronate Related Compound C
Synonym(s) [1-Hydroxy-2-(2-pyridinyl)ethylidene]bis(phosphonic acid), monohydrate [3,6-bis[(3-pyridinyl)methyl]-2,5-dihydroxy-2,5-dioxido-1,4,2,5-dioxadiphosphorinane-3,6-diyl]bis[phosphonic acid] disodium tetrahydrate salt [2-(3-pyridinyl)ethylidene-1,1]bis(phosphonic acid)
Environmentally Preferable
CAS No. 1220806-40-5 329003-65-8 75755-10-1
Formula C₇H₁₁NO₇P₂ C₁₄H₃₀N₂Na₂O₁₉P₄ C₇H₁₁NO₆P₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR