Filtration capsules, Sartopore® Platinum
Proveedor: Sartorius
5495307H8-OO-A 5495307H9-OO-A
SARP5495307H8-OO-AEA 1340 EUR
SARP5495307H8-OO-A SARP5495307H9-OO-A
Filtration capsules, Sartopore® Platinum
Filtros Cápsulas de filtro de membrana
Sartopore® Platinum sterilising grade MidiCaps® are self-contained sterile filter units designed for pharmaceutical and biotechnology industry. These filters contain a unique heterogeneous double layer of hydrophilised polyethersulfone membranes and are specially designed to meet the requirements for filtration of a broad range of pharmaceutical products.
  • Ready to use
  • Ideal for scale-up
  • Compatible with a pH range from 1 to 14 pH
  • Can be easily wetted with lowest flushing volume
PES membrane; support fleece, core and end caps: PP; O-rings: silicone;More Product Information
Order Now
ESPECIFICACIONES
Environmentally Preferable
Entrada/salida 1" Single-stepped hose barb
Ø×L 77×161 mm
Presión máx. 5 bar @ 20 °C; 3 @ 50 °C