VWR®, Precision Digital Waterproof HACCP Thermometers with PT1000 Probe
Proveedor: VWR Collection
620-2644EA 232 EUR
620-2644 620-2645 620-2646
VWR®, Precision Digital Waterproof HACCP Thermometers with PT1000 Probe
Termómetros Termómetros digitales
Accurate measurements for laboratories, quality management, and monitoring of production processes. Application: Food (HACCP), medical/pharmaceutical science, chemistry, aquaristics, fish farming, aquaculture, and more.
  • Easy handling
  • Battery life time >6000 hours
  • Device and probe robust and waterproof
  • With PT1000, two-wire, isolated steam- and waterproof, permanently connected to device
  • Response time T90: Approx. 10
  • Can be recalibrated at 0 °C and scale
Información accesorios: Compliance with HACCP and DIN EN 13485.
Información de suministro: Device incl. probe, battery, calibration protocol and manual.