Chemkey® Reagents
Proveedor: Hach
9425200
9429100
9429300
9429000
9429200
9429400
9429600
8499300
8499400
8636000
8636100
8636200
8636300
8636400
8636500
8636600
8791900
9429500
705-0952EA
82.2
EUR
705-0952
705-0945
705-0949
705-0943
705-0953
705-0947
705-0948
705-0944
705-0946
705-0954
705-0955
705-0956
705-0957
705-0958
705-0959
705-0960
HACH8791900
HACH9429500
Chemkey® Reagents
Kits para test medioambiente
Chemkey® reagents are a new technology that enable the SL1000 Portable Parallel Analyzer colorimeter to perform the same tests with less than half the manual steps.
- All chemicals and processes are entirely contained inside the Chemkey®
- There are no powder pillows or glass vials to handle
- Get highly accurate results with less opportunity for errors in a fraction of the time
There is no zeroing, no mixing, no shaking, no chemicals or vials to handle. All you have to do is insert the chemkey and let the PPA-SL1000 read you back the results.