Angelicin ≥98%, crystalline
Proveedor: Cayman Chemical
|
Atención
|
Sinónimos:
Furo[2,3-h]benzopyran-2-one, 2H-Furo[2,3-h]chromen-2-one, Isopsoralen, Bakuchicin, NSC 404563
11683-5
11683-10
11683-25
CAYM11683-5EA
143
EUR
CAYM11683-5
CAYM11683-10
CAYM11683-25
Angelicin ≥98%, crystalline
Angelicin
Angelicin is a furanocoumarin typically isolated from the seeds of P. corylifolia. Like other furanocoumarins, angelicin has antibacterial activities against a number of Gram (+) and Gram (–) bacteria. It has also been shown to prevent tacrine-induced cytotoxicity in human liver-derived HepG2 cells (EC₅₀=47 μg/ml) and vascular relaxation in phenylephrine-precontracted rat aorta. Angelicin also weakly inhibits topoisomerase II (IC₅₀=404 μM).
Formula:
C₁₁H₆O₃ MW: 186.17 g/mol Temperatura de almacenaje: Congelador |
Número MDL:
MFCD00064930 Núm. CAS: 523-50-2 |
Resultados test especificaciones
Purity | ≥98% |
Formulation | A crystalline solid |
λmax | 247, 299 nm |
Storage | –20 °C |
Stability | ≥2 years |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...