Cloxacillin sodium salt monohydrate
Proveedor: Thermo Scientific
|
Peligro
|
455300010
455300050
ACRO455300010EA
65.8
EUR
ACRO455300010
ACRO455300050
Cloxacillin sodium salt monohydrate
Cloxacillin sodium salt monohydrate
Formula:
C₁₉H₁₈ClN₃O₅S MW: 457,86 g/mol Temperatura de almacenaje: Nevera |
Número MDL:
MFCD00150735 Núm. CAS: 7081-44-9 |
Resultados test especificaciones
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Water | 3.0 to 5.0 % |
HPLC | ≥94 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...