Ethylenediaminetetraacetic acid disodium magnesium salt hydrate 97%
Proveedor: Thermo Fisher Scientific
Sinónimos:
Ethylenediaminetetraacetic acid disodium magnesium salt xhydrate, EDTA magnesium disodium salt hydrate, EDTA-Na2Mg salt
457930250
457931000
457935000
ACRO457930250EA
27.5
EUR
ACRO457930250
ACRO457931000
ACRO457935000
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate 97%
Ethylenediaminetetraacetic acid disodium magnesium salt hydrate
Formula:
C₁₀H₁₂MgN₂Na₂O₈·xH₂O Temperatura de almacenaje: Ambiente |
Número MDL:
MFCD00078217 Núm. CAS: 194491-32-2 |
Resultados test especificaciones
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Assay | ≥96.0 % (on dry substance) |
Loss on drying | 10 % to 20 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...