Quantus®, Cleanroom Shoe Covers
Proveedor: VWR Collection
QNTS106-6STWH00MIDEA 0 EUR
QNTS106-6STWH00MID QNTS106-6STWH00LID QNTS106-6STWH0XLID
Quantus®, Cleanroom Shoe Covers
Ropa de trabajo
These low shoe covers are made from Quantus® Standard fabric, a breathable, high-quality polyester fabric.
  • Elastic band at upper side
  • Press studs for width adjustment
  • Flexible plastic sole
  • Double reinforcement with fabric
Designed to meet the requirements of cleanroom class 100 (ISO 5).
Weight: 95 g/m²