Digestion units for Kjeldahl digestion, KJELDATHERM® KT/KT-L
Proveedor: GERHARDT
12-0700 12-0702 12-0704 12-0706 12-0725 12-0727 12-0729 12-0731 12-0743 12-0745
GERH12-0700EA 0 EUR
GERH12-0700 GERH12-0702 GERH12-0704 GERH12-0706 GERH12-0725 GERH12-0727 GERH12-0729 GERH12-0731 GERH12-0743 GERH12-0745
Digestion units for Kjeldahl digestion, KJELDATHERM® KT/KT-L
Digestores Estaciones de trabajo de mineralización
These high-precision, programmable digestion units are ideal for Kjeldahl digestion in 100, 250 or 400 ml digestion tubes. Depending on the sample volume, you can choose between 8-, 20- or 40- place systems.
  • Aluminium digestion block with optimum energy utilisation and effective all-round insulation
  • Sample tubes and extraction unit made of high quality DURAN® glass
  • Automatic control of TURBOSOG and cooling water valve
  • Illuminated digestion chamber with enamelled, acid-resistant surface
  • Pre-heating function for warming up the digestion block
  • Data
More Product Information
Order Now
ESPECIFICACIONES
Modelo KT 8s KT 8s-BS KT-L 8s KT-L 8s-BS KT 20s KT 20s-BS KT-L 20s KT-L 20s-BS KT 40s KT-L 40s
Environmentally Preferable
Nº de tubos 8 8 8 8 20 20 20 20 40 40
Potencia 1000 W 1000 W 1160 W 1160 W 2200 W 2200 W 2360 W 2360 W 2200 W 2360 W
Programas 99 99 99 99 99 99 99 99 99 99
Precisión de la temperatura ±5 °C ±5 °C ±5 °C ±5 °C ±5 °C ±5 °C ±5 °C ±5 °C ±5 °C ±5 °C
Intervalo de temperaturas Max. 430 °C Max. 430 °C Max. 430 °C Max. 430 °C Max. 430 °C Max. 430 °C Max. 430 °C Max. 430 °C Max. 430 °C Max. 430 °C
Ancho × Fondo × Alto 515×483×728 mm 515×483×728 mm 515×483×728 mm 515×483×728 mm 515×483×728 mm 515×483×728 mm 515×483×728 mm 515×483×728 mm 515×483×728 mm 515×483×728 mm
Peso 31 kg 31 kg 44 kg 44 kg 40 kg 40 kg 53 kg 53 kg 40 kg 53 kg