Lisinopril dihydrate 98.0-102% (base secada)
Proveedor: Thermo Fisher Scientific
|
Peligro
|
Sinónimos:
(S)-1-((S)-6-Amino-2-(((S)-1-carboxy-3-phenylpropyl)amino)hexanoyl)pyrrolidine-2-carboxylic acid dihydrate, 1-[Nalpha-[(S)-1-Carboxy-3-phenylpropyl]-L-lysyl]-L-proline
462610010
462610050
ACRO462610010EA
74.2
EUR
ACRO462610010
ACRO462610050
Lisinopril dihydrate 98.0-102% (base secada)
Lisinopril dihydrate
Formula:
C₂₁H₃₅N₃O₇ Pto. de fusión: 148 °C Temperatura de almacenaje: Nevera |
Número MDL:
MFCD01698825 Núm. CAS: 83915-83-7 |
Resultados test especificaciones
Appearance | White to almost white Crystalline powder |
Infrared spectrum | Conforms |
Water | 8 to 9.5 % |
Solubility | (1 % in water) Clear solution |
Residue after ignition | ≤0.1 % |
Titration with NaOH | 98.0 to 102.0 % (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...