Anti-BAX Rabbit Polyclonal Antibody

Proveedor: ENZO LIFE SCIENCES
ADIAAS040D ADIAAS040F
ENZOADIAAS040DEA 225 EUR
ENZOADIAAS040D ENZOADIAAS040F
Anti-BAX Rabbit Polyclonal Antibody
Anticuerpos
Bax is an appromimately 22 kDa pro-apoptotic protein belonging to the Bcl-2 family of proteins, which are characterized by the presence of at least one Bcl-2 homology (BH) domains. Bax normally resides in the cytoplasm, but following an apoptotic stimuli, Bax undergoes a conformational change and translocates to mitochondrial membranes where it inserts and mediates the release of cytochrome c. This activates caspase-3 and perpetuates the apoptotic cascade.

  • For Western Blot

Recognizes human, mouse, rat, bovine, monkey, pig and rabbit Bax. Detects a band of ~22kDa by Western blot.

Type: Primary
Antigen: BAX
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR