Anti-CRP55 Mouse Monoclonal Antibody

Proveedor: Enzo Life Sciences
ADISPA601D ADISPA601F
ENZOADISPA601DEA 306 EUR
ENZOADISPA601D ENZOADISPA601F
Anti-CRP55 Mouse Monoclonal Antibody
Anticuerpos
Calreticulin (CRT) is a multifunctional, multi-compartmental protein most abundant in the ER lumen. CRT contains the ER-retrieval sequence, KDEL, and has been best characterized as a soluble molecular chaperone of new or misfolded proteins and a Ca2+- binding protein. Both CRT and its membrane bound homolog, Calnexin (CNX) interact with proteins and glycoproteins that have monoglucosylated N-glycans. The CRT/CNX cycle promotes correct folding, inhibits aggregation of folding intermediates, blocks premature oligimerization, regulates ER degradation, and provides quality control by preventing incompletely folded glycoproteins from exiting to the Golgi complex.

  • For Immunocytochemistry, Immunoprecipitation, Western Blot

Recognizes human and monkey Calreticulin. Detects a band of ~63kDa by Western blot.

Type: Primary
Antigen: CRP55
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR