Anti-HSP70 Rabbit Polyclonal Antibody

Proveedor: ENZO LIFE SCIENCES
ADISPA812D ADISPA812F ADISPA812J
ENZOADISPA812DEA 223 EUR
ENZOADISPA812D ENZOADISPA812F ENZOADISPA812J
Anti-HSP70 Rabbit Polyclonal Antibody
Anticuerpos
The Hsp70 family of heat shock protiens contains multiple homologs ranging in size from 66-78 kDa, and are the eukaryotic equivalents of the bacterial DnaK. The most studied Hsp70 members include the cytosolic stress-induced Hsp70 (Hsp72), the constitutive cytosolic Hsc70 (Hsp73), and the ER-localized BiP (Grp78). Hsp70 family members contain highly conserved N-terminal ATP-ase and C-terminal protein binding domains. Binding of peptide to Hsp70 is assisted by Hsp40, and stimulates the inherent ATPase activity of Hsp70, facilitating ATP hydrolysis and enhanced peptide binding. Hsp70 nucleotide exchange and substrate binding coordinates the folding of newly synthesized proteins, the re-folding of misfolded or denatured proteins, coordinates trafficking of proteins across cellular membranes, inhibits protein aggregation, and targets the degradation of proteins via the proteasomal pathway.

  • For Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot

Recognizes human, mouse, rat, beluga, bovine, dog, Drosophila, fish, guinea pig, hamster, monkey, pig and sheep HSP70 (HSP72). Detects a band of ~70kDa by Western blot.

Type: Primary
Antigen: HSP70
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR