Anti-HSP65 Mouse Monoclonal Antibody

Proveedor: ENZO LIFE SCIENCES
ADISPA881D ADISPA881F
ENZOADISPA881DEA 295 EUR
ENZOADISPA881D ENZOADISPA881F
Anti-HSP65 Mouse Monoclonal Antibody
Anticuerpos
Hsp65 is a member of the Hsp60 family of heat shock proteins isolated from Mycobacterium bovis BCG. Hsp65 from M. bovis is identical to that of M. tuberculosis, and similar to that of M. leprae, two highly pathogenic strains of mycobacterium. Hsp65 is the immunodominant antigen during mycobacterial infection and vaccination, and has been linked to the development of autoimmune adjuvant arthritis in rats.

  • For Western Blot

Recognizes human, mouse, rat, bovine, bacteria (E. coli, M. leprae, M. bovis), cryptosporidium, Drosophila, hamster and guinea pig HSP65. Detects a band of ~65kDa by Western blot.

Type: Primary
Antigen: HSP65
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR