Anti-HSF1 Rat Monoclonal Antibody

Proveedor: ENZO LIFE SCIENCES
ADISPA950D ADISPA950F
ENZOADISPA950DEA 275 EUR
ENZOADISPA950D ENZOADISPA950F
Anti-HSF1 Rat Monoclonal Antibody
Anticuerpos
HSFs (Heat Shock family of transcription factors), which consists of HSF 1-4, bind to highly conserved Heat shock elements (HSEs) in the promoter regions of heat shock genes, ultimately regulating the expression of Heat shock proteins (Hsps). On exposure to heat shock and other stresses, HSF1 localizes within seconds to discrete nuclear granules and on recovery from stress, HSF1 rapidly dissipates from the stress granules to a diffuse nucleoplasmic distribution.

  • For Western Blot

Recognizes human, mouse, rat, monkey and rabbit HSF1. Predicted MW of ~60kDa. Detects a band of ~85kDa by Western blot.

Type: Primary
Antigen: HSF1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR