Anti-OPN Rabbit Polyclonal Antibody

Proveedor: ENZO LIFE SCIENCES
ALX210309R020 ALX210309R100
ENZOALX210309R020EA 273 EUR
ENZOALX210309R020 ENZOALX210309R100
Anti-OPN Rabbit Polyclonal Antibody
Anticuerpos
Osteopontin (OPN) is an acidic extracellular matrix cell adhesion protein that is relatively abundant not only in bone matrix, plasma, urine, and milk, but is also found in malignant and atherogenic tissues. Phosphorylation, glycosylation and calcium modifications allow intact and fragmented Osteopontin to direct a variety of diverse responses including tissue remodeling, inflammation and cell survival. Plasma Osteopontin has been shown to provide prognostic information in breast, prostate, colon, and lung cancers as well as metastatic carcinomas. The notable presence of Osteopontin in a variety of tumors is strongly correlated to pathological stage, suggesting its critical role in tumor invasiveness, progression and metastasis. In addition, Osteopontin inhibits inducible nitric oxide synthase activity, thereby protecting tumor cells from NO-mediated macrophage cytotoxic attack. Osteopontin is found in atherosclerotic plaques and may drive a number of diabetic vascular pathologies.

  • For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

Recognizes human osteopontin. Does not cross-react with human osteonectin or bone sialoprotein.

Reconstitute with 20µl or 100µl distilled water.

Type: Primary
Antigen: OPN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR