Anti-CD40L Hamster Monoclonal Antibody

Proveedor: ENZO LIFE SCIENCES
ALX804331C100 ALX804331C500
ENZOALX804331C100EA 395 EUR
ENZOALX804331C100 ENZOALX804331C500
Anti-CD40L Hamster Monoclonal Antibody
Anticuerpos
Host: Hamster, Isotype: IgG1

Recognizes mouse CD40L.

Type: Primary
Antigen: CD40L
Clonality: Monoclonal
Clone: 0
Conjugation:
Epitope:
Host: Hamster
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR