Anti-Ubiquitin Mouse Monoclonal Antibody
Proveedor: ENZO LIFE SCIENCES
BMLPW06000025 BMLPW06000100
ENZOBMLPW06000025EA 165 EUR
ENZOBMLPW06000025 ENZOBMLPW06000100
Anti-Ubiquitin Mouse Monoclonal Antibody
Anticuerpos
Host: Mouse, Isotype: IgG2a
  • For ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Recognizes K63-linked polyubiquitin in wide range of species. Does not cross-react with any other isopeptide-linked (K6, K11, K27, K29, K33, or K48) polyubiquitinylated species.
Type: Primary
Antigen: UBIQUITIN
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Species Independent