Anti-INOS Rabbit Polyclonal Antibody

Proveedor: ENZO LIFE SCIENCES
BMLSA2000025 BMLSA2000100
ENZOBMLSA2000025EA 209 EUR
ENZOBMLSA2000025 ENZOBMLSA2000100
Anti-INOS Rabbit Polyclonal Antibody
Anticuerpos
Host: Rabbit

  • For Immunocytochemistry, Immunohistochemistry, Western Blot

Recognizes human, rat and mouse iNOS. Detects a band of ~130kDa by Western blot. Does not cross-react with eNOS or nNOS.

Type: Primary
Antigen: INOS
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

ESPECIFICACIONES

Simbolo antígeno INOS INOS
Nombre del antígeno Inducible Nitric Oxide Synthase Inducible Nitric Oxide Synthase
Environmentally Preferable
Sinónimos de antígenos macrophage NOS|inducible nitric oxide synthase|nitric oxide synthase 2|inducible NO synthase|inducible NOS|MAC-NOS|inducible|NOS type II|macrophage|nitric oxide synthase macrophage NOS|inducible nitric oxide synthase|nitric oxide synthase 2|inducible NO synthase|inducible NOS|MAC-NOS|inducible|NOS type II|macrophage|nitric oxide synthase
Tipo de anticuerpo Primario Primario
Clonalidad Polyclonal Polyclonal
Reactividad Human
Huésped Rabbit Rabbit
Isotipo
Western blot Yes Yes
InmunoQuímica Yes Yes
Forma Liquid, neat serum Liquid, neat serum

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR