Anti-EIF2 alpha Rabbit Polyclonal Antibody

Proveedor: ENZO LIFE SCIENCES
BMLSA4050020 BMLSA4050100
ENZOBMLSA4050020EA 195 EUR
ENZOBMLSA4050020 ENZOBMLSA4050100
Anti-EIF2 alpha Rabbit Polyclonal Antibody
Anticuerpos
Host: Rabbit

  • For Western Blot

Recognizes human and yeast eIF2alpha phosphorylated at Ser51 and mouse eIF2alpha phosphorylated at Ser52. Does not recognize unphosphorylated form of eIF2alpha.

Type: Primary
Antigen: EIF2 alpha
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR