Anti-P53 Mouse Monoclonal Antibody [clone: PAb122]

Proveedor: ENZO LIFE SCIENCES
ADIKAMCC002D ADIKAMCC002F
ENZOADIKAMCC002DEA 261 EUR
ENZOADIKAMCC002D ENZOADIKAMCC002F
Anti-P53 Mouse Monoclonal Antibody [clone: PAb122]
Anticuerpos
Type: Primary
Antigen: P53
Clonality: Monoclonal
Clone: PAb122
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR