Anti-HA-tag Mouse Monoclonal Antibody [clone: 16B12]

Proveedor: ENZO LIFE SCIENCES
ENZABS1200200 ENZABS1200500 ENZABS1201000
ENZOENZABS1200200EA 583 EUR
ENZOENZABS1200200 ENZOENZABS1200500 ENZOENZABS1201000
Anti-HA-tag Mouse Monoclonal Antibody [clone: 16B12]
Anticuerpos
Recommended Applications: IF, IHC, IP, WB

Species reactivity:

Type: Primary
Antigen: HA-tag
Clonality: Monoclonal
Clone: 16B12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Hamster, Cat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR