Anti-TAU Mouse Monoclonal Antibody [clone: C3]

Proveedor: ENZO LIFE SCIENCES
ENZABS2140500 ENZABS2141000
ENZOENZABS2140500EA 1370 EUR
ENZOENZABS2140500 ENZOENZABS2141000
Anti-TAU Mouse Monoclonal Antibody [clone: C3]
Anticuerpos
Recommended Applications: ELISA, IHC, WB

Species reactivity: Human

Type: Primary
Antigen: TAU
Clonality: Monoclonal
Clone: C3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Sheep
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Más About VWR