Anti-SNCA Mouse Monoclonal Antibody [clone: LB509]
Proveedor: ENZO LIFE SCIENCES
ENZABS2180200 ENZABS2180500
ENZOENZABS2180200EA 891 EUR
ENZOENZABS2180200 ENZOENZABS2180500
Anti-SNCA Mouse Monoclonal Antibody [clone: LB509]
Anticuerpos
Recommended Applications: ELISA, IHC, WB
Species reactivity: Human
Type: Primary
Antigen: SNCA
Clonality: Monoclonal
Clone: LB509
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Xenopus, Mammalian, Chicken