CD40 Soluble (Human) ELISA kit
Proveedor: Enzo Life Sciences
ENZOALX850262KI01EA 996.6 EUR
ENZOALX850262KI01 ALEXALX850262KI01
CD40 Soluble (Human) ELISA kit
Ensayos ELISAs
For the quantitative determination of human soluble CD40 in cell culture supernatants, serum and other body fluids.
  • Assay Time: <3,25 hours