ADMA direct (mouse/rat) ELISA kit
Proveedor: Enzo Life Sciences
ALEXALX850327KI01EA 1810 EUR
ALEXALX850327KI01 ENZOALX850327KI01
ADMA direct (mouse/rat) ELISA kit
Ensayos ELISAs
For the quantitative detection of asymmetrical dimethylarginine (ADMA) in mouse or rat EDTA-plasma and serum. Does not cross-react with SDMA, NMMA or L-arginine.