Annexin V-FITC apoptosis detection kit
Proveedor: ENZO LIFE SCIENCES
ALX850020KI01 ALX850020KI02
ENZOALX850020KI01EA 860 EUR
ENZOALX850020KI01 ENZOALX850020KI02
Annexin V-FITC apoptosis detection kit
Ensayos Ensayos celulares
For the detection of apoptotic cells by flow cytometry.
FITC-labelled recombinant human annexin V shows bright green fluorescence (Ex(max): 488 nm, Em(max): 530 nm).
Información de suministro: Kit contains Annexin V-FITC, binding buffer and propidium iodide.
New Product
Order Now
ESPECIFICACIONES
Environmentally Preferable
Aplicación Flow Cytometry, Fluorescence Microscopy, Fluorescent Detection Flow Cytometry, Fluorescence Microscopy, Fluorescent Detection
Tamaño 300 tests 100 tests
Pureza ≥98% (SDS-PAGE; HPLC) ≥98% (SDS-PAGE; HPLC)
Condiciones de envío 4 °C 4 °C
Condiciones de almacenamiento 4 °C 4 °C